accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 1731-84-6

IUPAC Name: methyl nonanoate
Synonyms: methyl nonanoate (ester)
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: IJXHLVMUNBOGRR-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)OC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)