missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nonanoate 99.5+%, TCI America™

Catalog No. S03075ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03075ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03075ML Supplier TCI America Supplier No. S03075ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1731-84-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00009569
InChI Key IJXHLVMUNBOGRR-UHFFFAOYSA-N
Synonym methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
PubChem CID 15606
ChEBI CHEBI:44499
IUPAC Name methyl nonanoate
SMILES CCCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 214°C
Quantity 5 mL
Formula Weight 172.27
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonanoate [Standard Material for GC]
EINECSNumber (2)-0782&(2)-0798
RTECSNumber RA6906000
TSCA Yes