missing translation for 'onlineSavingsMsg'
Learn More

Methyl nonanoate, 95%, Thermo Scientific™

Catalog No. p-3586452
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC385560050 5g Glass bottle
AC385560250 25g Glass bottle
AC385561000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC385560050 Supplier Thermo Scientific Chemicals Supplier No. 385560050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1731-84-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009569
InChI Key IJXHLVMUNBOGRR-UHFFFAOYSA-N
Synonym methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
PubChem CID 15606
ChEBI CHEBI:44499
IUPAC Name methyl nonanoate
SMILES CCCCCCCCC(=O)OC

Specifications

Density 0.8740g/mL
Boiling Point 213°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4210 to 1.423
Linear Formula CH3(CH2)7C(O)OCH3
Quantity 5g
Specific Gravity 0.874
Solubility Information Solubility in water: practically insoluble
Formula Weight 172.27
Percent Purity 95%
Chemical Name or Material Methyl nonanoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-052-7

RUO – Research Use Only