missing translation for 'onlineSavingsMsg'
Learn More

Methyl Nonanoate 96.0+%, TCI America™

Catalog No. N029025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N029025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N029025ML Supplier TCI America Supplier No. N029025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1731-84-6
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00009569
InChI Key IJXHLVMUNBOGRR-UHFFFAOYSA-N
Synonym methyl pelargonate, pelargonic acid methyl ester, methyl nonylate, nonanoic acid, methyl ester, methyl n-nonanoate, nonanoic acid methyl ester, unii-7xkd6qoh68, fema no. 2724, methyl ester nonanoic acid, 7xkd6qoh68
PubChem CID 15606
ChEBI CHEBI:44499
IUPAC Name methyl nonanoate
SMILES CCCCCCCCC(=O)OC

Specifications

Color Colorless
Boiling Point 214°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonanoate
EINECSNumber (2)-0782&(2)-0798
RTECSNumber RA6906000
TSCA Yes