CAS RN 1619-37-0
IUPAC Name:
(5-phenyl-1,2-oxazol-3-yl)methanol
Synonyms:
(5-phenyl-1,2-oxazol-3-yl)methanol
Molecular Weight (g/mol):
175.19
Molecular Formula:
C10H9NO2
InChi Key:
BPAFLGGUEBMWRN-UHFFFAOYSA-N
SMILES:
OCC1=NOC(=C1)C1=CC=CC=C1
1
–
4
of
4
results
2
(5-Phenylisoxazol-3-yl)methanol, 97%, Thermo Scientific™
CAS: 1619-37-0 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.187 MDL Number: MFCD01444104 InChI Key: BPAFLGGUEBMWRN-UHFFFAOYSA-N PubChem CID: 1481082 IUPAC Name: (5-phenyl-1,2-oxazol-3-yl)methanol SMILES: C1=CC=C(C=C1)C2=CC(=NO2)CO
4
(5-Phenylisoxazol-3-yl)methanol 98.0+%, TCI America™
CAS: 1619-37-0 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.187 MDL Number: MFCD01444104 InChI Key: BPAFLGGUEBMWRN-UHFFFAOYSA-N PubChem CID: 1481082 IUPAC Name: (5-phenyl-1,2-oxazol-3-yl)methanol SMILES: C1=CC=C(C=C1)C2=CC(=NO2)CO