missing translation for 'onlineSavingsMsg'
Learn More

(5-Phénylisoxazol-3-yl)méthanol, 97%, Thermo Scientific™

Numéro de catalogue. CC30509DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC30509DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC30509DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC30509DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 1619-37-0
Formule moléculaire C10H9NO2
Poids moléculaire (g/mol) 175.187
Numéro MDL MFCD01444104
Clé InChI BPAFLGGUEBMWRN-UHFFFAOYSA-N
PubChem CID 1481082
Nom de l’IUPAC (5-phényl-1,2-oxazol-3-yl)méthanol
SOURIRES C1=CC=C(C=C1)C2=CC(=NO2)CO

Spécifications

Nom ou substance chimique (5-Phenylisoxazol-3-yl)methanol
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Synonyme 5-phenylisoxazol-3-yl methanol, 5-phenyl-3-isoxazolyl methanol, 5-phenyl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol, 5-phenyl, 3-hydroxymethyl-5-phenylisoxazole, 5-phenylisoxazol-3-yl methan-1-ol, 5-phenylisoxazole-3-methanol, 3-isoxazolemethanol,5-phenyl, 5-phenyl-3-hydroxymethylisoxazole, 5-phenyl-isoxazol-3-yl methanol
Poids de formule 175.19
Pourcentage de pureté 97%
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.