missing translation for 'onlineSavingsMsg'
Learn More

(5-Phenylisoxazol-3-yl)methanol 98.0+%, TCI America™

Catalog No. P23865G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23865G 5 g
P23861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23865G Supplier TCI America Supplier No. P23865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1619-37-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD01444104
InChI Key BPAFLGGUEBMWRN-UHFFFAOYSA-N
PubChem CID 1481082
IUPAC Name (5-phenyl-1,2-oxazol-3-yl)methanol
SMILES C1=CC=C(C=C1)C2=CC(=NO2)CO

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Synonym 5-phenylisoxazol-3-yl methanol, 5-phenyl-3-isoxazolyl methanol, 5-phenyl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol, 5-phenyl, 3-hydroxymethyl-5-phenylisoxazole, 5-phenylisoxazol-3-yl methan-1-ol, 5-phenylisoxazole-3-methanol, 3-isoxazolemethanol,5-phenyl, 5-phenyl-3-hydroxymethylisoxazole, 5-phenyl-isoxazol-3-yl methanol
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (5-Phenylisoxazol-3-yl)methanol