CAS RN 1617-90-9
IUPAC Name:
methyl (15S,17S,19S)-15-ethyl-17-hydroxy-1,11-diazapentacyclo[9.6.2.0²,⁷.0⁸,¹⁸.0¹⁵,¹⁹]nonadeca-2,4,6,8(18)-tetraene-17-carboxylate
Synonyms:
minorin vincachron arteriovinca vinodrel retard vinkametrin vincamidol pervincamine vincaminum vincamine vincamina
Molecular Weight (g/mol):
354.45
Molecular Formula:
C21H26N2O3
InChi Key:
RXPRRQLKFXBCSJ-GIVPXCGWSA-N
SMILES:
CC[C@@]12CCCN3CCC4=C([C@H]13)N(C1=CC=CC=C41)[C@](O)(C2)C(=O)OC
1
–
2
of
2
results
1
Thermo Scientific Chemicals Vincamine, 98%
CAS: 1617-90-9 Molecular Formula: C21H26N2O3
2
Vincamine 98.0+%, TCI America™
CAS: 1617-90-9 Molecular Formula: C21H26N2O3 Molecular Weight (g/mol): 354.45 MDL Number: MFCD00078054 InChI Key: RXPRRQLKFXBCSJ-GIVPXCGWSA-N PubChem CID: 15376 ChEBI: CHEBI:9985 SMILES: CCC12CCCN3C1C4=C(CC3)C5=CC=CC=C5N4C(C2)(C(=O)OC)O