missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vincamine, 98%

Catalog No. AAJ6054506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6054506 5 g
AAJ6054503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6054506 Supplier Thermo Scientific Chemicals Supplier No. J6054506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A vasodilator | CAS: 1617-90-9 | C21H26N2O3, C21H26N2O3 | 354.45 g/mol

A vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1617-90-9
Molecular Formula C21H26N2O3

Specifications

Quantity 5 g
Description White to Pale Yellow
Content And Storage 2°C to 8°C
For Use With (Application) A vasodilator
Formula Weight 354.45g/mol
Melting Point 232°C
Product Type Signal Transduction Reagent
Form Powder
Assay 98%

RUO – Research Use Only