missing translation for 'onlineSavingsMsg'
Learn More

Vincamine 98.0+%, TCI America™

Catalog No. V00615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00615G 5 g
V00611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V00615G Supplier TCI America Supplier No. V00615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1617-90-9
Molecular Formula C21H26N2O3
Molecular Weight (g/mol) 354.45
MDL Number MFCD00078054
InChI Key RXPRRQLKFXBCSJ-GIVPXCGWSA-N
PubChem CID 15376
ChEBI CHEBI:9985
SMILES CCC12CCCN3C1C4=C(CC3)C5=CC=CC=C5N4C(C2)(C(=O)OC)O

Specifications

Melting Point 232°C
Color White
Quantity 5 g
Formula Weight 354.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Vincamine
RTECSNumber YY8575000
TSCA No