accessibility menu, dialog, popup

UserName

CAS RN 1617-53-4

IUPAC Name: 8-[5-(5,7-dihydroxy-4-oxo-4H-chromen-2-yl)-2-hydroxyphenyl]-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Synonyms: amentoflavone 3',8''-biapigenin
Molecular Weight (g/mol): 538.46
Molecular Formula: C30H18O10
InChi Key: YUSWMAULDXZHPY-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C(=C2O1)C1=CC(=CC=C1O)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
Grade 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)