missing translation for 'onlineSavingsMsg'
Learn More

Amentoflavone, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013287
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013287 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013287 Supplier Merck Emd Millipore Supplier No. 1857110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Amentoflavone is a flavonoid which is used as an antiviral agent.

Featured Industries: Food and Beverages

Specifications

CAS 1617-53-4
Physical Form Neat
Linear Formula C30H18O10
MDL Number MFCD00017470
Synonym Didemethyl-ginkgetin; I3′,II8-Biapigenin; Tridemethylsciadopitysin
Type Analytical Standard
Quantity 10 mg
Formula Weight 538.46
Percent Purity ≥98% (HPLC)
Grade Analytical Standard