missing translation for 'onlineSavingsMsg'
Learn More

Amentoflavone, Thermo Scientific™

Catalog No. AC469071000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469071000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC469071000 Supplier Thermo Scientific Chemicals Supplier No. 469071000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1617-53-4
Molecular Formula C30 H18 O10

Specifications

Quantity 100 mg
Formula Weight 538.48
Physical Form Powder
Color White to Yellow to Beige
Purity ≥97.5%
Chemical Name or Material Amentoflavone
Hazard Category Exclamation mark
Recommended Storage Normal conditions