accessibility menu, dialog, popup

UserName

CAS RN 1603-91-4

IUPAC Name: 4-methyl-1,3-thiazol-2-amine
Synonyms: 2-thiazolamine, 4-methyl-
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: OUQMXTJYCAJLGO-UHFFFAOYSA-N
SMILES: CC1=CSC(N)=N1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)