missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylthiazole, 98+%

Catalog No. AAA1484722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1484722 100 g
AAA1484706 5 g
AAA1484714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1484722 Supplier Thermo Scientific Chemicals Supplier No. A1484722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1603-91-4
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD00005329
InChI Key OUQMXTJYCAJLGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiazole, 4-methylthiazol-2-amine, 2-thiazolamine, 4-methyl, 2-amino-4-methylthiazol, 4-methyl-2-thiazolamine, 4-methyl-2-thiazolylamine, 2-amino-4-methyl-1,3-thiazole, normotiroide, thiazole, 2-amino-4-methyl, 4-methyl-2-aminothiazole
PubChem CID 74143
ChEBI CHEBI:39753
IUPAC Name 4-methyl-1,3-thiazol-2-amine
SMILES CC1=CSC(=N1)N

Specifications

Melting Point 42°C to 46°C
Boiling Point 231°C to 233°C
Flash Point 107°C (224°F)
Odor Characteristic
Quantity 100 g
Beilstein 107890
Sensitivity Hygroscopic
Merck Index 14,450
Formula Weight 114.17
Percent Purity ≥98%
Chemical Name or Material 2-Amino-4-methylthiazole
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-505-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only