accessibility menu, dialog, popup

UserName

CAS RN 15708-41-5

IUPAC Name: iron(3+) sodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: iron(3+) sodium ion(4-) edta
Molecular Weight (g/mol): 367.05
Molecular Formula: C10H12FeN2NaO8
InChi Key: MKWYFZFMAMBPQK-UHFFFAOYSA-J
SMILES: [Na+].[Fe+3].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Physical Form 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)