missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iron EDTA Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018857
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018857 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018857 Supplier Merck Emd Millipore Supplier No. PHR16451G

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 15708-41-5
Linear Formula C10H12N2 NaFeO8 · 3H2 O
Synonym Ethylenediaminetetraacetic acid iron(III) sodium salt trihydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 421.09
Grade Certified Reference Material