missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA, Sodium Ferric Salt, MP Biomedicals
Specifications
Specifications
| Quantity | 50 g |
| Formula Weight | 367.05 |
| Grade | Not Available |
| Physical Form | Powder |
| Chemical Name or Material | EDTA |