accessibility menu, dialog, popup

UserName

OONO

CAS RN 154447-36-6

IUPAC Name: 2-(morpholin-4-yl)-8-phenyl-4H-chromen-4-one
Synonyms: 2-(morpholin-4-yl)-8-phenylchromen-4-one
Molecular Weight (g/mol): 307.35
Molecular Formula: C19H17NO3
InChi Key: CZQHHVNHHHRRDU-UHFFFAOYSA-N
SMILES: O=C1C=C(OC2=C(C=CC=C12)C1=CC=CC=C1)N1CCOCC1
Form 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results