missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholino-8-phenylchromone 98.0+%, TCI America™

Catalog No. M241025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M241025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M241025MG Supplier TCI America Supplier No. M241025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Morpholino-substituted chromone compound used as a laboratory research reagent.

Chemical Identifiers

CAS 154447-36-6
Molecular Formula C19H17NO3
Molecular Weight (g/mol) 307.349
MDL Number MFCD00270881
InChI Key CZQHHVNHHHRRDU-UHFFFAOYSA-N
Synonym 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran, LY 294002
PubChem CID 3973
ChEBI CHEBI:65329
IUPAC Name 2-morpholin-4-yl-8-phenylchromen-4-one
SMILES C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 184°C
Color White
Quantity 25 mg
Formula Weight 307.35
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-Morpholino-8-phenylchromone
TSCA No
Recommended Storage Freezer