missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals LY 294002, 98%

Catalog No. AAJ67289MB
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67289MB 25 mg
AAJ67289LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67289MB Supplier Thermo Scientific Chemicals Supplier No. J67289MB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 154447-36-6 | C19H17NO3, C19H17NO3 | 307.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154447-36-6
Molecular Formula C19H17NO3
Synonym 2-(4-morpholinyl)-8-phenyl-4H-1-benzopyran-4-one;

Specifications

Quantity 25 mg
Description White to pale yellow
Content And Storage -30°C to -10°C
Formula Weight 307.35g/mol
Form solid
Assay 98%

RUO – Research Use Only