accessibility menu, dialog, popup

UserName

CAS RN 153766-81-5

IUPAC Name: potassium trifluoro(phenyl)boranuide
Synonyms: potassium trifluoro(phenyl)boranuide
Molecular Weight (g/mol): 184.01
Molecular Formula: C6H5BF3K
InChi Key: DVAFPKUGAUFBTJ-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)