missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phenyltrifluoroborate 98.0+%, TCI America™

Catalog No. P15825G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15825G 5 g
P15821G 1 g
P158225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P15825G Supplier TCI America Supplier No. P15825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 153766-81-5
Molecular Formula C6H5BF3K
Molecular Weight (g/mol) 184.01
MDL Number MFCD01318172
InChI Key DVAFPKUGAUFBTJ-UHFFFAOYSA-N
Synonym potassium phenyltrifluoroborate, potassium trifluoro phenyl borate, potassium trifluoro phenyl boranuide, pubchem11425, potassiumphenyltrifluoroborate, amtb100, phenyltrifluoropotassioboron v, potassium trifluoro phenyl borate 1-, potassium ion trifluoro phenyl boranuide, potassium tris fluoranyl-phenyl-boranuide
PubChem CID 23675248
IUPAC Name potassium;trifluoro(phenyl)boranuide
SMILES [B-](C1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Melting Point 301°C
Color White
Quantity 5 g
Formula Weight 184.01
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Phenyltrifluoroborate