accessibility menu, dialog, popup

UserName

CAS RN 151767-02-1

IUPAC Name: sodium 2-[1-({[(1R)-1-{3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanyl}methyl)cyclopropyl]acetate
Synonyms: singulair montelukast sodium salt singular montair montelukast sodium
Molecular Weight (g/mol): 608.17
Molecular Formula: C35H35ClNNaO3S
InChi Key: LBFBRXGCXUHRJY-HKHDRNBDSA-M
SMILES: [Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)