missing translation for 'onlineSavingsMsg'
Learn More

Montelukast Sodium Hydrate 98.0+%, TCI America™

Catalog No. M2340200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2340200MG 200 mg
M23401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2340200MG Supplier TCI America Supplier No. M2340200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 151767-02-1
Molecular Formula C35H35ClNNaO3S
Molecular Weight (g/mol) 608.17
MDL Number MFCD00931431
InChI Key LBFBRXGCXUHRJY-HKHDRNBDSA-M
Synonym montelukast sodium, singulair, montair, unii-u1o3j18sfl, montelukast sodium usan, montelukast na, montelukast sodium salt, u1o3j18sfl, singulair tn
PubChem CID 23663996
ChEBI CHEBI:6993
IUPAC Name sodium 2-[1-({[(1R)-1-{3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanyl}methyl)cyclopropyl]acetate
SMILES [Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1

Specifications

Melting Point 115°C
Color White
pH 9.7 (1% aq. soln.)
Quantity 200 mg
Formula Weight 608.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Montelukast Sodium Hydrate
RTECSNumber GZ0698000
TSCA No