missing translation for 'onlineSavingsMsg'
Learn More

Montelukast Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013205
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013205 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013205 Supplier Merck Emd Millipore Supplier No. PHR16031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Montelukast Sodium is an orally administered leukotriene receptor antagonist drug that acts by inhibiting the cysteinyl leukotriene (CysLT1) receptor. It is effective against leukotrienes responsible for bronchoconstriction and inflammation of the airways leading to asthma problems.

Specifications

CAS 151767-02-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C35H35 ClNO3 S·Na
Synonym Montelukast sodium
Type Certified Reference Material
Quantity 1 g
Formula Weight 608.17
Grade Certified Reference Material