accessibility menu, dialog, popup

UserName

H3CSBrBr

CAS RN 149703-84-4

IUPAC Name: 2,5-dibromo-3-octylthiophene
Synonyms: 2,5-dibromo-3-octylthiophene
Molecular Weight (g/mol): 354.14
Molecular Formula: C12H18Br2S
InChi Key: PJGBSUPPENVFAD-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=C(Br)SC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results