missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-n-octylthiophene 98.0+%, TCI America™

Catalog No. D40535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40535G Supplier TCI America Supplier No. D40535G
Only null left

Chemical Identifiers

CAS 149703-84-4
Molecular Formula C12H18Br2S
Molecular Weight (g/mol) 354.14
MDL Number MFCD00672013
InChI Key PJGBSUPPENVFAD-UHFFFAOYSA-N
Synonym 2,5-dibromo-3-n-octylthiophene, 2,5-dibromo-3-octyl-thiophene, acmc-1c21j, thiophene, 2,5-dibromo-3-octyl, 2,5-bis bromanyl-3-octyl-thiophene, 2,5-dibromo-3-octylthiophene
PubChem CID 4385790
IUPAC Name 2,5-dibromo-3-octylthiophene
SMILES CCCCCCCCC1=C(Br)SC(Br)=C1

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 g
Formula Weight 354.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-n-octylthiophene
TSCA No
Recommended Storage Refrigerator