accessibility menu, dialog, popup

UserName

CAS RN 14898-67-0

IUPAC Name: ruthenium(3+) trichloride
Synonyms: ruthenium(III) chloride
Molecular Weight (g/mol): 207.42
Molecular Formula: Cl3Ru
InChi Key: YBCAZPLXEGKKFM-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Ru+3]
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)