missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) chloride hydrate, Premion™, 99.99% (metals basis)

Catalog No. AA4336409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4336409 10 g
AA4336404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4336409 Supplier Thermo Scientific Chemicals Supplier No. 04336409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14898-67-0 | Cl3Ru | 207.42 g/mol

Ruthenium(III) chloride hydrate is used as a starting material in ruthenium chemistry. It acts as a precursor to many ruthenium compounds, which finds application as catalysts in asymmetric hydrogenation and alkene metathesis. It serves as a catalyst in the preparation of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine. Further, it is used in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst for the efficient dehydrogenation of arylmethyl alcohols to the corresponding aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14898-67-0
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.42
MDL Number MFCD00149844
InChI Key YBCAZPLXEGKKFM-UHFFFAOYSA-K
IUPAC Name ruthenium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ru+3]

Specifications

Melting Point 100°C (decomposition)
Color Black
Physical Form Crystalline
Quantity 10 g
UN Number UN3260
Merck Index 14,8302
Solubility Information Soluble in water.
Formula Weight 207.43 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ruthenium (III) chloride hydrate
Show More Show Less
Hazard Category H290-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H290-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 233-167-5
RTECSNumber VM2650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only