CAS RN 148493-34-9
IUPAC Name:
(2,6-dichloropyridin-3-yl)boronic acid
Synonyms:
2,6-dichloropyridin-3-ylboronic acid
Molecular Weight (g/mol):
191.8
Molecular Formula:
C5H4BCl2NO2
InChi Key:
XBBLBQZAVMHEER-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(Cl)N=C1Cl
1
–
2
of
2
results
1
Promotions Available
2,6-Dichloropyridine-3-boronic acid, 95%
2,6-Dichloropyridine-3-boronic acid, 95%
CAS: 148493-34-9 Molecular Formula: C5H4BCl2NO2 Molecular Weight (g/mol): 191.802 MDL Number: MFCD03094989 InChI Key: XBBLBQZAVMHEER-UHFFFAOYSA-N Synonym: 2,6-dichloropyridine-3-boronic acid,2,6-dichloropyridin-3-yl boronic acid,2,6-dichloro-3-pyridineboronic acid,3-borono-2,6-dichloropyridine,2,6-dichloropyridin-3-yl-3-boronic acid,2,6-dichloro-3-pyridinyl-boronic acid,boronic acid, 2,6-dichloro-3-pyridinyl,2,6-dichloropyridine-3-boronicacid,pubchem5039,acmc-209d0p PubChem CID: 2762712 IUPAC Name: (2,6-dichloropyridin-3-yl)boronic acid SMILES: B(C1=C(N=C(C=C1)Cl)Cl)(O)O