missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D47071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D47071G Supplier TCI America Supplier No. D47071G

Specifications

Chemical Name or Material 2,6-Dichloropyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 148493-34-9
Melting Point 150°C
Color White-Yellow
Molecular Formula C5H4BCl2NO2
Quantity 1 g
MDL Number MFCD03094989
Synonym 2,6-dichloropyridine-3-boronic acid, 2,6-dichloropyridin-3-yl boronic acid, 2,6-dichloro-3-pyridineboronic acid, 3-borono-2,6-dichloropyridine, 2,6-dichloropyridin-3-yl-3-boronic acid, 2,6-dichloro-3-pyridinyl-boronic acid, boronic acid, 2,6-dichloro-3-pyridinyl, 2,6-dichloropyridine-3-boronicacid, pubchem5039, acmc-209d0p
InChI Key XBBLBQZAVMHEER-UHFFFAOYSA-N
SMILES B(C1=C(N=C(C=C1)Cl)Cl)(O)O
IUPAC Name (2,6-dichloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 191.802
PubChem CID 2762712
Formula Weight 191.80
Physical Form Crystalline Powder
Show More Show Less