missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-3-boronic acid, 95%

Catalog No. AAH27406MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27406MD 250 mg
AAH2740603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27406MD Supplier Thermo Scientific Chemicals Supplier No. H27406MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 148493-34-9 | C5H4BCl2NO2 | 191.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148493-34-9
Molecular Formula C5H4BCl2NO2
Molecular Weight (g/mol) 191.802
MDL Number MFCD03094989
InChI Key XBBLBQZAVMHEER-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine-3-boronic acid, 2,6-dichloropyridin-3-yl boronic acid, 2,6-dichloro-3-pyridineboronic acid, 3-borono-2,6-dichloropyridine, 2,6-dichloropyridin-3-yl-3-boronic acid, 2,6-dichloro-3-pyridinyl-boronic acid, boronic acid, 2,6-dichloro-3-pyridinyl, 2,6-dichloropyridine-3-boronicacid, pubchem5039, acmc-209d0p
PubChem CID 2762712
IUPAC Name (2,6-dichloropyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)Cl)Cl)(O)O

Specifications

Quantity 250 mg
Formula Weight 191.81
Percent Purity 95%
Chemical Name or Material 2,6-Dichloropyridine-3-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only