accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 14660-52-7

IUPAC Name: ethyl 5-bromopentanoate
Synonyms: ethyl 5-bromopentanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: AFRWBGJRWRHQOV-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCBr
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)