missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Bromovalerate 97.0+%, TCI America™

Catalog No. B155725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B155725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B155725G Supplier TCI America Supplier No. B155725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
Synonym ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr

Specifications

Color Yellow
Boiling Point 227°C
Quantity 25 g
Formula Weight 209.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 5-Bromovalerate