accessibility menu, dialog, popup

UserName

H3CCH3ONSOHOH3CN

CAS RN 144060-53-7

IUPAC Name: 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
Synonyms: febuxostat
Molecular Weight (g/mol): 316.38
Molecular Formula: C16H16N2O3S
InChi Key: BQSJTQLCZDPROO-UHFFFAOYSA-N
SMILES: CC(C)COC1=CC=C(C=C1C#N)C1=NC(C)=C(S1)C(O)=O
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)