missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Febuxostat, 98%

Catalog No. AC461580050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461580050 5 g
AC461580010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461580050 Supplier Thermo Scientific Chemicals Supplier No. 461580050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 144060-53-7 | C16H16N2O3S | 316.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 144060-53-7
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00871598
InChI Key BQSJTQLCZDPROO-UHFFFAOYSA-N
IUPAC Name 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC(C)COC1=CC=C(C=C1C#N)C1=NC(C)=C(S1)C(O)=O

Specifications

Melting Point 209.0°C
Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Formula Weight 316.38
Chemical Name or Material Febuxostat

RUO – Research Use Only