accessibility menu, dialog, popup

UserName

CAS RN 141460-19-7

HHHHNOONOONOONOOCNSCNSRu

CAS RN 141460-19-7

IUPAC Name: λ²-ruthenium(2+) tetrahydrogen bis(carbothioylazanide) bis([2,2'-bipyridine]-4,4'-dicarboxylate)
Synonyms: λ²-ruthenium(2+) tetrahydrogen bis(carbothioylazanide) bis([2,2'-bipyridine]-4,4'-dicarboxylate)
Molecular Weight (g/mol): 705.64
Molecular Formula: C26H16N6O8RuS2
InChi Key: VMISXESAJBVFNH-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[H+].[Ru++].[N-]=C=S.[N-]=C=S.[O-]C(=O)C1=CC(=NC=C1)C1=NC=CC(=C1)C([O-])=O.[O-]C(=O)C1=CC(=NC=C1)C1=NC=CC(=C1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results