missing translation for 'onlineSavingsMsg'
Learn More

Bis(isothiocyanato)bis(2,2'-bipyridyl-4,4'-dicarboxylato)ruthenium(II) 85.0+%, TCI America™

Catalog No. B4372200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4372200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4372200MG Supplier TCI America Supplier No. B4372200MG
Only null left

Chemical Identifiers

CAS 141460-19-7
Molecular Formula C26H16N6O8RuS2
Molecular Weight (g/mol) 705.638
MDL Number MFCD11042213
InChI Key VMISXESAJBVFNH-UHFFFAOYSA-N
Synonym N3 Dye
PubChem CID 121235232
IUPAC Name 2-(4-carboxylatopyridin-2-yl)pyridine-4-carboxylate;hydron;ruthenium(2+);diisothiocyanate
SMILES [H+].[H+].[H+].[H+].C1=CN=C(C=C1C(=O)[O-])C2=NC=CC(=C2)C(=O)[O-].C1=CN=C(C=C1C(=O)[O-])C2=NC=CC(=C2)C(=O)[O-].C(=[N-])=S.C(=[N-])=S.[Ru+2]

Specifications

Color Black-Purple
Quantity 200 mg
Formula Weight 705.64
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(isothiocyanato)bis(2,2′-bipyridyl-4,4′-dicarboxylato)ruthenium(II)