CAS RN 1405-41-0
Gentamycin Sulfate (White Powder), Fisher BioReagents
CAS: 1405-41-0 Molecular Formula: C21H43N5O7·H2SO4 Synonym: Gentiomycin C,Garamycin
Thermo Scientific Chemicals Gentamicin sulfate
CAS: 1405-41-0 Synonym: Garamycin,Gentamicin Sulfate
Gentamicin sulfate, 600 I.U./mg
CAS: 1405-41-0 MDL Number: MFCD00270181
Thermo Scientific Chemicals Gentamycin Sulfate
CAS: 1405-41-0 Molecular Formula: C21H43N5O7·H2SO4 MDL Number: MFCD00270181 Synonym: Garamycin,Gentamicin Sulfate
Thermo Scientific Chemicals Gentamicin sulfate, 20 mg/ml in distilled water, sterile-filtered
CAS: 1405-41-0 MDL Number: MFCD00270181
Gentamicin Sulfate, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.
Gentamicin Sulfate, MP Biomedicals™
CAS: 1405-41-0 Synonym: Garamycin, Genoptic, GM Sulfate;Gentiomycin C
Gentamicin Sulfate, MP Biomedicals™
CAS: 1405-41-0 Synonym: Garamycin, Gentamicin sulfate salt, Gentiomycin C
Gentamycin Sulfate, MP Biomedicals
CAS: 1405-41-0
Gentamycin Sulfate, USP, MP Biomedicals™
CAS: 1405-41-0 Synonym: Gentamycin Sulfate
Gentamicin Sulfate USP Crystalline MP Biomedicals
CAS: 1405-41-0 Molecular Formula: C21H43N5O7 Synonym: Gentamycin Sulfate
MP Biomedicals™ Gentamicin Sulfate, 50 mg/mL
Effective against gram-positive and gram-negative bacteria, as well as mycoplasma.
- 1
- 2