missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Gentamicin Sulfate, 50 mg/mL
Description
Gentamicin is an aminoglycoside antibiotic complex that inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms, including strains resistant to tetracycline, chloramphenicol, kanamycin, and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus, and Streptococcus. It is effective against mycoplasma as well.
- Used as a selection agent (gentamicin-resistance gene) in molecular biology applications.
- Key applications: cell culture applications, antimicrobial treatment.
Specifications
Specifications
| Physical Form | Liquid |
| CAS | 1405-41-0, 7732-18-5 |
| pH | 4 to 6 |
| Sterility | Sterile |
| Color | Clear Colorless to Pale Yellow |
| Concentration | 50 mg/mL |
| Quantity | 1 x 10 mL |
| Synonym | Garamycin, Gentamicin sulfate salt, Gentiomycin C |
| Grade | Not Available |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.