CAS RN 136779-28-7
IUPAC Name:
1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane
Synonyms:
1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane
Molecular Weight (g/mol):
362.48
Molecular Formula:
C22H36P2
InChi Key:
GVVCHDNSTMEUCS-UHFFFAOYNA-N
SMILES:
CCC1CCC(CC)P1C1=CC=CC=C1P1C(CC)CCC1CC
1
–
1
of
1
results
1
Promotions Available
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+%
(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+%
CAS: 136779-28-7 Molecular Formula: C22H36P2 Molecular Weight (g/mol): 362.478 MDL Number: MFCD00142321 InChI Key: GVVCHDNSTMEUCS-MUGJNUQGSA-N Synonym: ethyl-duphos, s,s,unii-fri8026kma,s,s-et-duphos,fri8026kma,+-1,2-bis 2s,5s-2,5-diethylphospholano benzene,+-duphos,2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane,s,s-ethyl-duphos PubChem CID: 5702640 IUPAC Name: (2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane SMILES: CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC