missing translation for 'onlineSavingsMsg'
Learn More

(+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene, 97+%

Catalog No. AAH60981MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60981MD 250 mg
AAH6098103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60981MD Supplier Thermo Scientific Chemicals Supplier No. H60981MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136779-28-7 | C22H36P2 | 362.478 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 136779-28-7
Molecular Formula C22H36P2
Molecular Weight (g/mol) 362.478
MDL Number MFCD00142321
InChI Key GVVCHDNSTMEUCS-MUGJNUQGSA-N
Synonym ethyl-duphos, s,s, unii-fri8026kma, s,s-et-duphos, fri8026kma, +-1,2-bis 2s,5s-2,5-diethylphospholano benzene, +-duphos, 2s,5s-1-2-2s,5s-2,5-diethylphospholan-1-yl phenyl-2,5-diethylphospholane, s,s-ethyl-duphos
PubChem CID 5702640
IUPAC Name (2S,5S)-1-[2-[(2S,5S)-2,5-diethylphospholan-1-yl]phenyl]-2,5-diethylphospholane
SMILES CCC1CCC(P1C2=CC=CC=C2P3C(CCC3CC)CC)CC

Specifications

Density 1.01
Refractive Index 1.581
Quantity 250 mg
Beilstein 4814174
Sensitivity Air Sensitive
Optical Rotation +265° (c=1 in Hexane)
Formula Weight 362.47
Percent Purity ≥97%
Chemical Name or Material (+)-1,2-Bis[(2S,5S)-2,5-diethyl-1-phospholanyl]benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only