accessibility menu, dialog, popup

UserName

CAS RN 136466-94-9

IUPAC Name: (2,6-difluoropyridin-3-yl)boronic acid
Synonyms: 2,6-difluoropyridin-3-ylboronic acid
Molecular Weight (g/mol): 158.9
Molecular Formula: C5H4BF2NO2
InChi Key: LCCZTROJRJFXNV-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)N=C(F)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)