missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-pyridineboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D40061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40061G Supplier TCI America Supplier No. D40061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2,6-Difluoro-3-pyridineboronic Acid (contains varying amounts of Anhydride)
CAS 136466-94-9
Color White-Yellow
Molecular Formula C5H4BF2NO2
Quantity 1 g
MDL Number MFCD03788245
Synonym 2,6-difluoropyridine-3-boronic acid, 2,6-difluoropyridin-3-yl boronic acid, 2,6-difluoro-3-pyridineboronic acid, 2,6-difluoropyridin-3-yl-3-boronic acid, 2,6-difluoropyridine-3-boronicacid, 2,6-difluoropyridin-3-ylboronicacid, 2,6-difluoro-3-pyridyl boronic acid, 2,6-difluoro-3-pyridylboronic acid, boronic acid, 2,6-difluoro-3-pyridinyl, 2,6-difluoro-3-pyridinyl boronic acid
InChI Key LCCZTROJRJFXNV-UHFFFAOYSA-N
SMILES B(C1=C(N=C(C=C1)F)F)(O)O
IUPAC Name (2,6-difluoropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 158.899
PubChem CID 2762713
Formula Weight 158.90
Percent Purity 97.0 to 113.0%
Physical Form Crystalline Powder
Show More Show Less