missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoropyridine-3-boronic acid, 95%, Thermo Scientific™

Catalog No. AAH2750903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2750903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2750903 Supplier Thermo Scientific Chemicals Supplier No. H2750903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 136466-94-9
Molecular Formula C5H4BF2NO2
Molecular Weight (g/mol) 158.899
MDL Number MFCD03788245
InChI Key LCCZTROJRJFXNV-UHFFFAOYSA-N
Synonym 2,6-difluoropyridine-3-boronic acid, 2,6-difluoropyridin-3-yl boronic acid, 2,6-difluoro-3-pyridineboronic acid, 2,6-difluoropyridin-3-yl-3-boronic acid, 2,6-difluoropyridine-3-boronicacid, 2,6-difluoropyridin-3-ylboronicacid, 2,6-difluoro-3-pyridyl boronic acid, 2,6-difluoro-3-pyridylboronic acid, boronic acid, 2,6-difluoro-3-pyridinyl, 2,6-difluoro-3-pyridinyl boronic acid
PubChem CID 2762713
IUPAC Name (2,6-difluoropyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)F)F)(O)O

Specifications

Quantity 1 g
Formula Weight 158.9
Percent Purity 95%
Chemical Name or Material 2,6-Difluoropyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures