accessibility menu, dialog, popup

UserName

CAS RN 13609-67-1

IUPAC Name: (1R,3aS,3bS,9aR,9bS,10S,11aS)-10-hydroxy-1-(2-hydroxyacetyl)-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl butanoate
Synonyms: hydrocortisone 17-butyrate hydrocortisone-17α-butyrate cortisol 17-butyrate hydrocortisone butyrate (usan:ban:jan)
Molecular Weight (g/mol): 432.56
Molecular Formula: C25H36O6
InChi Key: BMCQMVFGOVHVNG-TUFAYURCSA-N
SMILES: CCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)