accessibility menu, dialog, popup

UserName

CAS RN 130693-82-2

IUPAC Name: hydrogen (2R,4R)-4-(ethylamino)-2-methyl-1,1-dioxo-2H,3H,4H-1λ⁶-thieno[2,3-b]thiopyran-6-sulfonamide chloride
Synonyms: dorzolamide
Molecular Weight (g/mol): 360.89
Molecular Formula: C10H17ClN2O4S3
InChi Key: OSRUSFPMRGDLAG-CIRBGYJCSA-N
SMILES: [H+].[Cl-].CCN[C@@H]1C[C@@H](C)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results