missing translation for 'onlineSavingsMsg'
Learn More

Dorzolamide Hydrochloride 98.0+%, TCI America™

Catalog No. D4189200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4189200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4189200MG Supplier TCI America Supplier No. D4189200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130693-82-2
Molecular Formula C10H17ClN2O4S3
Molecular Weight (g/mol) 360.89
MDL Number MFCD00884659
InChI Key OSRUSFPMRGDLAG-CIRBGYJCSA-N
PubChem CID 6918132
ChEBI CHEBI:4703
IUPAC Name hydrogen (2R,4R)-4-(ethylamino)-2-methyl-1,1-dioxo-2H,3H,4H-1λ⁶-thieno[2,3-b]thiopyran-6-sulfonamide chloride
SMILES [H+].[Cl-].CCN[C@@H]1C[C@@H](C)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O

Specifications

Melting Point 281°C
Color White-Yellow
Quantity 200 mg
Formula Weight 360.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dorzolamide Hydrochloride
RTECSNumber XJ9095163
TSCA No