missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dorzolamide hydrochloride

Catalog No. AC466612500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466612500 250 mg
AC466610010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466612500 Supplier Thermo Scientific Chemicals Supplier No. 466612500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130693-82-2
Molecular Formula C10H16N2O4S3·HCl

Specifications

Quantity 250 mg
Solubility Solubility aq. soln.: soluble aq. soln.. Other solubilities: slightly soluble in methanol
Melting Point 281.0°C
Formula Weight 360.89
Chemical Name or Material Dorzolamide hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only